siRNA Details
Gene Symbol: ISM1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040043.1 | 2 | 317 | |||
GenBank mRNA | BC038745.1 | - | 2 | 304 | ||
DB453950.1 | - | 2 | 317 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ISM1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040043.1 | 2 | 317 | |||
GenBank mRNA | BC038745.1 | - | 2 | 304 | ||
DB453950.1 | - | 2 | 317 |