siRNA Details
Gene Symbol: MEIS1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046625.1 | 2 | 223 | |||
GenBank mRNA | CA772438.1 | - | 2 | 488 |

More Information
Other Species Targeted


UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MEIS1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046625.1 | 2 | 223 | |||
GenBank mRNA | CA772438.1 | - | 2 | 488 |