siRNA Details
Gene Symbol: DICER1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015415.1 | 4 | 759 | |||
GenBank mRNA | AK127179.1 | - | 2 | 2289 | ||
BI754942.1 | - | 4 | 685 | |||
BX648898.1 | - | 1 | 6135 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DICER1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015415.1 | 4 | 759 | |||
GenBank mRNA | AK127179.1 | - | 2 | 2289 | ||
BI754942.1 | - | 4 | 685 | |||
BX648898.1 | - | 1 | 6135 |