siRNA Details
Gene Symbol: PCOLCE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038910.1 | 2 | 283 | |||
GenBank mRNA | AK097705.1 | - | 2 | 283 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB047350.1 | - | 2 | 283 | Chr. 7 - 100602795 - 100604038 [-] on Build GRCh38 |
