siRNA Details
Gene Symbol: MAMDC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040076.1 | 4 | 1425 | |||
GenBank mRNA | BC043510.1 | - | 4 | 1369 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MAMDC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040076.1 | 4 | 1425 | |||
GenBank mRNA | BC043510.1 | - | 4 | 1369 |