siRNA Details
Gene Symbol: TOB1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038458.1 | 1 | 479 | |||
GenBank mRNA | BC039664.1 | - | 1 | 344 | ||
BP421960.1 | - | 1 | 14 | |||
BQ222352.1 | - | 1 | 489 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TOB1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038458.1 | 1 | 479 | |||
GenBank mRNA | BC039664.1 | - | 1 | 344 | ||
BP421960.1 | - | 1 | 14 | |||
BQ222352.1 | - | 1 | 489 |