siRNA Details
Gene Symbol: KCTD21-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102280.1 | 1 | 269 | |||
NR_102281.1 | 1 | 269 | ||||
GenBank mRNA | DA156609.1 | - | 1 | 253 | ||
DA737651.1 | - | 1 | 220 | |||
DA814941.1 | - | 1 | 269 | |||
DB082989.1 | - | 1 | 268 |
