siRNA Details
Gene Symbol: BOLA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045634.1 | 2 | 797 | |||
NR_045636.1 | 3 | 671 | ||||
NR_045637.1 | 2 | 601 | ||||
GenBank mRNA | AK123453.1 | - | 3 | 607 | ||
BC030102.2 | - | 2 | 546 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG706622.1 | - | 2 | 551 | Chr. 2 - 74148023 - 74149056 [+] on Build GRCh38 |
