siRNA Details
Gene Symbol: NCAM1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034101.1 | 1, 2 | 277 | |||
GenBank mRNA | AK293718.1 | - | 1, 2 | 208 | ||
DC305174.1 | - | 1, 2 | 128 | |||
DC309426.1 | - | 1, 2 | 277 |
