siRNA Details
Gene Symbol: LINC00963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038955.1 | 2 | 661 | |||
GenBank mRNA | AK024177.1 | - | 2 | 214 | ||
AK092686.1 | - | 2 | 181 | |||
BC038559.1 | - | 3 | 354 | |||
DR006520.1 | - | 2 | 667 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU122149.1 | - | 2 | 214 | Chr. 9 - 129489107 - 129502906 [+] on Build GRCh38 |
