siRNA Details
Gene Symbol: UCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015379.3 | 3 | 1519 | |||
GenBank mRNA | AK092401.1 | - | 3 | 1505 | ||
GU357550.1 | - | 3 | 1371 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX346962.1 | - | 1 | 761 | Chr. 19 - 15834765 - 15835610 [+] on Build GRCh38 |
