siRNA Details
Gene Symbol: LINC01504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110952.1 | 2 | 206 | |||
NR_110953.1 | 2 | 206 | ||||
GenBank mRNA | AA436218.1 | - | 2 | 124 | ||
AK095210.1 | - | 2 | 206 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110952.1 | 2 | 206 | |||
NR_110953.1 | 2 | 206 | ||||
GenBank mRNA | AA436218.1 | - | 2 | 124 | ||
AK095210.1 | - | 2 | 206 |