siRNA Details
Gene Symbol: TTTY16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001552.1 | 1, 2 | 63 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF527828.3 | - | 1, 2 | 63 | Chr. Y - 7699357 - 7701247 [-] on Build GRCh38 |
