siRNA Details
Gene Symbol: ALMS1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046762.1 | 2 | 780 | |||
GenBank mRNA | BC041487.1 | - | 2 | 787 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ALMS1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046762.1 | 2 | 780 | |||
GenBank mRNA | BC041487.1 | - | 2 | 787 |