siRNA Details
Gene Symbol: LOC102723505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110876.1 | 1, 2 | 93 | |||
GenBank mRNA | BG182087.1 | - | 1, 2 | 98 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG207469.1 | - | 119 | Chr. - - [-] on Build |
