siRNA Details
Gene Symbol: LOC101928226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125950.1 | 1 | 81 | |||
GenBank mRNA | DY654363.1 | - | 1 | 81 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DY654617.1 | - | 93 | Chr. - - [-] on Build |
