siRNA Details
Gene Symbol: PSMG3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021487.2 | 4, 5 | 816 | |||
NR_027329.1 | 4, 5 | 562 | ||||
GenBank mRNA | BC036405.1 | - | 4, 5 | 624 | ||
BC047649.1 | - | 4, 5 | 619 | |||
BX538028.1 | - | 4, 5 | 500 | |||
DA928000.1 | - | 4, 5 | 435 |
