siRNA Details
Gene Symbol: LINC01093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039975.1 | 1 | 100 | |||
NR_039976.1 | 1 | 100 | ||||
NR_039977.1 | 1 | 100 | ||||
GenBank mRNA | AY429594.1 | - | 1 | 100 | ||
AY429595.1 | - | 1 | 100 | |||
CB153796.1 | - | 1 | 90 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG568575.1 | - | 1 | 94 | Chr. 4 - 184893901 - 184899454 [-] on Build GRCh38 |
