siRNA Details
Gene Symbol: DSCAM-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038896.1 | 1 | 306 | |||
NR_038898.1 | 1 | 306 | ||||
NR_038899.1 | 1 | 305 | ||||
NR_038900.1 | 1 | 306 | ||||
GenBank mRNA | AF401029.1 | - | 1 | 306 | ||
AF401030.1 | - | 1 | 306 | |||
AF401031.1 | - | 1 | 306 | |||
AF401032.1 | - | 1 | 306 | |||
AF401033.1 | - | 1 | 306 | |||
AF401034.1 | - | 1 | 306 | |||
AF401035.1 | - | 1 | 305 |
