siRNA Details
Gene Symbol: DSCAM-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038896.1 | 3 | 1531 | |||
NR_038898.1 | 2 | 1173 | ||||
NR_038899.1 | 3 | 1075 | ||||
NR_038900.1 | 2 | 1044 | ||||
GenBank mRNA | AF401029.1 | - | 2 | 1173 | ||
AF401030.1 | - | 2 | 1044 | |||
AF401033.1 | - | 3 | 1525 | |||
AF401035.1 | - | 3 | 1075 |
