siRNA Details
Gene Symbol: C21orf62-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024622.1 | 4 | 511 | |||
NR_024623.1 | 4 | 499 | ||||
GenBank mRNA | BC034794.1 | - | 2 | 225 | ||
BC117397.1 | - | 4 | 393 | |||
BC117399.1 | - | 4 | 405 |
