siRNA Details
Gene Symbol: GDNF-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278094.1 | NP_001265023.1 | 1 | 812 | ||
NR_103441.1 | 3 | 380 | ||||
NR_103442.1 | 2 | 2689 | ||||
XM_017008916.1 | XP_016864405.1 | 4 | 1361 | |||
XM_017008917.1 | XP_016864406.1 | 3 | 1669 | |||
XM_017008918.1 | XP_016864407.1 | 4 | 1658 | |||
GenBank mRNA | DA359107.1 | - | 1 | 518 | ||
JF824129.1 | - | 1 | 812 | |||
JF824130.1 | - | 3 | 380 | |||
JF824131.1 | - | 2 | 2690 |
