siRNA Details
Gene Symbol: FAM182A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026713.1 | 5 | 1411 | |||
GenBank mRNA | AK095693.1 | - | 5 | 1370 | ||
BC131537.1 | - | 5 | 1363 |
Gene Symbol: FAM182B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026714.2 | 4 | 1783 | |||
GenBank mRNA | AK054844.1 | - | 1 | 148 |
