siRNA Details
Gene Symbol: C22orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302819.1 | NP_001289748.1 | 2 | 1235 | ||
NM_001302820.1 | NP_001289749.1 | 2 | 1162 | |||
NM_015372.2 | NP_056187.1 | 3 | 1026 | |||
NR_126507.1 | 3 | 953 | ||||
GenBank mRNA | AL050256.1 | - | 3 | 1014 |
