siRNA Details
Gene Symbol: SNHG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038110.1 | 1, 2 | 86 | |||
NR_038111.1 | 1, 2 | 86 | ||||
GenBank mRNA | AB073602.1 | - | 1, 2 | 75 | ||
AK129904.1 | - | 1, 2 | 82 | |||
BC042949.2 | - | 1 | 72 | |||
BX647993.1 | - | 1, 2 | 45 |
