siRNA Details
Gene Symbol: SNHG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038108.1 | 4 | 759 | |||
NR_038109.1 | 4 | 755 | ||||
NR_038110.1 | 3 | 660 | ||||
NR_038111.1 | 3 | 656 | ||||
GenBank mRNA | AB073602.1 | - | 3 | 645 | ||
AB447886.1 | - | 4 | 754 | |||
AK024090.1 | - | 1 | 125 | |||
AK055935.1 | - | 4 | 713 | |||
AK129904.1 | - | 3 | 652 | |||
BC042949.2 | - | 3 | 1828 | |||
BC100293.1 | - | 4 | 687 | |||
BC112893.1 | - | 1 | 458 | |||
BX647993.1 | - | 3 | 615 |
