siRNA Details
Gene Symbol: C10orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039380.3 | NP_001034469.2 | 2 | 2053 | ||
XM_017015895.1 | XP_016871384.1 | 3 | 1184 | |||
GenBank mRNA | AK055129.1 | - | 2 | 2031 | ||
AK311351.1 | - | 371 |
