siRNA Details
Gene Symbol: ZNF667-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036521.1 | 2 | 779 | |||
NR_036522.1 | 2 | 424 | ||||
GenBank mRNA | BC110411.1 | - | 2 | 522 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF667-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036521.1 | 2 | 779 | |||
NR_036522.1 | 2 | 424 | ||||
GenBank mRNA | BC110411.1 | - | 2 | 522 |