siRNA Details
Gene Symbol: ZNF667-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036521.1 | 2 | 595 | |||
NR_036522.1 | 2 | 240 | ||||
GenBank mRNA | BC008651.1 | - | 2 | 181 | ||
BC110411.1 | - | 2 | 338 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF667-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036521.1 | 2 | 595 | |||
NR_036522.1 | 2 | 240 | ||||
GenBank mRNA | BC008651.1 | - | 2 | 181 | ||
BC110411.1 | - | 2 | 338 |