siRNA Details
Gene Symbol: LOC93622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015433.1 | 2 | 1520 | |||
GenBank mRNA | AK026375.1 | - | 1 | 1824 | ||
BC006130.2 | - | 2 | 1520 | |||
BC021542.1 | - | 1 | 1796 | |||
BC026970.1 | - | 2 | 1526 | |||
BC040997.1 | - | 2 | 2167 | |||
BC066911.1 | - | 1 | 1904 |
