siRNA Details
Gene Symbol: CCDC13-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046732.1 | 1 | 73 | |||
GenBank mRNA | AK098828.1 | - | 1 | 85 | ||
DB179860.1 | - | 1 | 75 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC13-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046732.1 | 1 | 73 | |||
GenBank mRNA | AK098828.1 | - | 1 | 85 | ||
DB179860.1 | - | 1 | 75 |