siRNA Details
Gene Symbol: LINC00476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023389.1 | 1 | 760 | |||
GenBank mRNA | AF170307.1 | - | 1 | 263 | ||
AF194971.1 | - | 1 | 759 | |||
AK092137.1 | - | 1 | 373 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK818715.1 | - | 1 | 16 | Chr. 9 - 95870133 - 95875233 [-] on Build GRCh38 |
