siRNA Details
Gene Symbol: C15orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302797.1 | NP_001289726.1 | 2 | 816 | ||
NM_207445.3 | NP_997328.1 | 2 | 927 | |||
GenBank mRNA | AK096850.1 | - | 2 | 878 | ||
BC137300.1 | - | 2 | 830 | 9021297 | ||
BC137301.1 | - | 2 | 830 | 9021298 | ||
BC171789.1 | - | 2 | 720 | 9054443 | ||
BX647708.1 | - | 2 | 914 |
