siRNA Details
Gene Symbol: MRVI1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034093.2 | 1 | 295 | |||
NR_034094.2 | 1 | 295 | ||||
NR_046374.1 | 1 | 295 | ||||
NR_046375.1 | 1 | 295 | ||||
GenBank mRNA | BC028123.1 | - | 1 | 259 | 5167345 | |
BI520309.1 | - | 1 | 234 | |||
BU569149.1 | - | 1 | 222 | |||
BU570065.1 | - | 1 | 203 | |||
DB071465.1 | - | 1 | 298 | |||
DB454301.1 | - | 1 | 248 |
