siRNA Details
Gene Symbol: LINC00469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027146.1 | 1 | 43 | |||
GenBank mRNA | AK097638.1 | - | 1 | 43 | ||
BC101213.1 | - | 1 | 26 | |||
BC101214.1 | - | 1 | 26 | |||
BC101215.1 | - | 1 | 26 |
