siRNA Details
Gene Symbol: LOC648987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034086.1 | 2 | 2048 | |||
NR_104649.1 | 3 | 2164 | ||||
NR_104650.1 | 2 | 418 | ||||
NR_104651.1 | 3 | 523 | ||||
GenBank mRNA | BC013653.1 | - | 2 | 1962 | ||
BC039088.1 | - | 3 | 2164 | |||
BC046188.1 | - | 2 | 1961 | |||
HY041253.1 | - | 2 | 420 |
