siRNA Details
Gene Symbol: EIF3J-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034170.1 | 1 | 700 | |||
NR_034171.1 | 2 | 235 | ||||
GenBank mRNA | AK092547.1 | - | 1 | 451 | ||
BC026043.1 | - | 1 | 688 | |||
BC042851.1 | - | 1 | 454 | |||
BC066970.1 | - | 1 | 700 | |||
DB480720.1 | - | 2 | 235 |
