siRNA Details
Gene Symbol: WDFY3-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015359.1 | 4 | 1455 | |||
GenBank mRNA | AK124663.1 | - | 1 | 456 | ||
AY250185.1 | - | 7 | 1978 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WDFY3-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015359.1 | 4 | 1455 | |||
GenBank mRNA | AK124663.1 | - | 1 | 456 | ||
AY250185.1 | - | 7 | 1978 |