siRNA Details
Gene Symbol: FER1L6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040044.1 | 6 | 1587 | |||
GenBank mRNA | AK093040.1 | - | 6 | 1587 | ||
BC137357.1 | - | 6 | 1554 | 9021355 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FER1L6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040044.1 | 6 | 1587 | |||
GenBank mRNA | AK093040.1 | - | 6 | 1587 | ||
BC137357.1 | - | 6 | 1554 | 9021355 |