siRNA Details
Gene Symbol: LINC00301 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026946.1 | 7 | 841 | |||
GenBank mRNA | BC029583.1 | - | 6 | 724 | ||
BX103095.1 | - | 6 | 727 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00301 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026946.1 | 7 | 841 | |||
GenBank mRNA | BC029583.1 | - | 6 | 724 | ||
BX103095.1 | - | 6 | 727 |