siRNA Details
Gene Symbol: TP53TG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015381.1 | 2 | 531 | |||
GenBank mRNA | AB007455.1 | - | 2 | 554 | ||
AB007456.1 | - | 3 | 483 | |||
AB007457.1 | - | 2 | 334 | |||
AB007458.1 | - | 2 | 263 | |||
AK292782.1 | - | 2 | 548 | |||
BC002709.2 | - | 2 | 531 | |||
BC061927.1 | - | 2 | 484 | |||
BC103631.1 | - | 2 | 238 |
