siRNA Details
Gene Symbol: SPACA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316972.1 | NP_001303901.1 | 1 | 134 | ||
NR_024330.1 | 1 | 138 | ||||
XM_017026299.1 | XP_016881788.1 | 1 | 1657 | |||
GenBank mRNA | AY358799.1 | - | 1 | 138 | ||
BC104208.2 | - | 1 | 117 | 40031428 | ||
BC104209.2 | - | 1 | 117 |
