siRNA Details
Gene Symbol: LINC00518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027793.1 | 2 | 327 | |||
GenBank mRNA | BC016666.1 | - | 2 | 327 | ||
BC017496.1 | - | 2 | 46 | 3921932 | ||
BC028118.1 | - | 2 | 57 | 3922009 |
