siRNA Details
Gene Symbol: KTN1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027123.1 | 3 | 501 | |||
GenBank mRNA | BX248296.1 | - | 3 | 483 | ||
JX046911.1 | - | 3 | 506 | |||
JX628584.1 | - | 3 | 506 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KTN1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027123.1 | 3 | 501 | |||
GenBank mRNA | BX248296.1 | - | 3 | 483 | ||
JX046911.1 | - | 3 | 506 | |||
JX628584.1 | - | 3 | 506 |