siRNA Details
Gene Symbol: PAX8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015377.2 | 6 | 1720 | |||
GenBank mRNA | AK001856.1 | - | 3 | 868 | ||
AK056355.1 | - | 6 | 1358 | |||
AK307781.1 | - | 5 | 1068 | |||
AL390179.1 | - | 3 | 878 | |||
BC033562.1 | - | 5 | 1603 | |||
BC036699.2 | - | 6 | 1720 | |||
BX537688.1 | - | 5 | 1261 |
