siRNA Details
Gene Symbol: SPATA41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028139.1 | 1 | 1291 | |||
GenBank mRNA | AK057536.1 | - | 1 | 228 | ||
AK124283.1 | - | 1 | 1286 | |||
AY605659.1 | - | 1 | 544 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SPATA41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028139.1 | 1 | 1291 | |||
GenBank mRNA | AK057536.1 | - | 1 | 228 | ||
AK124283.1 | - | 1 | 1286 | |||
AY605659.1 | - | 1 | 544 |