siRNA Details
Gene Symbol: LOC401320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038889.1 | 1 | 50 | |||
GenBank mRNA | AK096056.1 | - | 1 | 30 | ||
AW952614.1 | - | 1 | 50 |
Gene Symbol: GLYATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545358.2 | XP_011543660.1 | 1 | 627 | ||
XM_011545359.2 | XP_011543661.1 | 1 | 628 |
