siRNA Details
Gene Symbol: LOC401320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038889.1 | 3 | 345 | |||
GenBank mRNA | AK027340.1 | - | 3 | 1652 | ||
AK096056.1 | - | 3 | 325 | |||
AK096687.1 | - | 3 | 269 | |||
BC016976.1 | - | 2 | 156 |
