siRNA Details
Gene Symbol: LINC00173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027346.1 | 1, 2 | 178 | |||
GenBank mRNA | BC038547.1 | - | 1, 2 | 178 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI837301.1 | - | 1, 2 | 181 | Chr. 12 - 116533447 - 116536513 [+] on Build GRCh38 |
