siRNA Details
Gene Symbol: SCOC-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033939.1 | 2 | 157 | |||
GenBank mRNA | BU932771.1 | - | 2 | 160 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA070842.1 | - | 2 | 168 | Chr. 4 - 140285188 - 140373403 [-] on Build GRCh38 |
